missing translation for 'onlineSavingsMsg'
Learn More

Tosyl Cyanide (>90%), TRC

Product Code. 30456914
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Unit Size:
2.5g
5g
Product Code. Quantity unitSize
30456914 2.5 g 2.5g
30450554 5 g 5g
30449879 25 g -
3 options
This item is not returnable. View return policy

Product Code. 30456914

Brand: Toronto Research Chemicals TRCT6310002.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19158-51-1
Formula Weight 181.0197
InChI Formula InChI=1S/C8H7NO2S/c1-7-2-4-8(5-3-7)12(10,11)6-9/h2-5H,1H3
IUPAC Name (4-methylphenyl)sulfonylformonitrile
Molecular Formula C8H7NO2S
Molecular Weight (g/mol) 181.21
SMILES Cc1ccc(cc1)S(=O)(=O)C#N
Synonym 4-Methylbenzenesulfonyl Cyanide, p-Toluenesulfonyl Cyanide, 4-Toluenesulfonyl Cyanide, Tosyl Cyanide, p-Tolylsulfonyl Cyanide
Chemical Name or Material Tosyl Cyanide

Specifications

Quantity 2.5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.