Learn More
Palintest™ Total Free Chlorine Test
Providing an inexpensive approach to water testing for applications such as aquarium monitoring and domestic and professional water treatment control, the kits are ideal for inclusion with such systems or for marketing to users of such equipment.
Brand:  Palintest™ PK011
Additional Details : Weight : 0.80000kg
Product Code. 11948936
  • 52.95€ / Pack of 50
Estimated Shipment: 10-05-2024
to see stock.
Add to basket
Description
Description
  • Packaged in a convenient box, Pocket Kits contain reagents and all accessories required to perform simple visual testing of parameters ranging from ammonia to zinc
  • Providing an inexpensive approach to water testing for applications such as aquarium monitoring and domestic and professional water treatment control 
  • Pocket Kits are all based on tablet reagents and visual color comparison or color/turbidity change methods
  • These economically priced kits will be of particular interest to companies marketing water treatment systems to domestic or industrial users
  • The kits are ideal for inclusion with such systems or for marketing to users of such equipment
Specifications
Specifications
50 Pack
0 to 2mg/L
50 Tests
Total Free Chlorine Test