Learn More
Macherey-Nagel™ Total Iron 100 Test Strip
Shop All Macherey-Nagel Products
Click to view available options
Quantity:
100 Pack
Unit Size:
Pack of 100
Description
Semi-quantitative test strips for the determination of Iron. Ideal for rapid and easy analysis of Iron directly on-site. No maintenance or calibration is required.
Order Info
Graduations
Specifications
Specifications
| Format | Semi Quantitative Test Strips |
| For Use With (Application) | Quick and easy determination of total iron |
| Graduations | 0 to 100mg/L |
| Quantity | 100 Pack |
| Type | Total Iron 100 Test Strip |