Få mere at vide
Få mere at vide
Aalborg™ Totalizer with Screen
Klik for at se tilgængelige muligheder
Width (Metric):
19.1 mm
Pakningsstørrelse:
Each
Beskrivelse
- Displays instantaneous, total and accumulated flow rates.
- Built-in Flow Linearizer (10 point linearization of the flow curve).
- Up to 47 different volumetric and mass flow engineering units.
- Compact design for unit mount, panel mount, wall mount or field mount applications.
- User-programmable, optically-isolated pulse output.
- Two programmable, optically-isolated, digital outputs.
- Flow controllers, set point command control via local LCD or digital interface.
- Programmable set point table with ramping up/down capability up to 16 steps.
- Free Configuration and Monitoring Utility software
Tekniske data
Tekniske data
| Display Type | LCD |
| Voltage | 0/5 V |
| Type | Totalizer |
| Height (Metric) | 76.2 mm |
| Width (Metric) | 19.1 mm |
| Electrical Requirements | 0 to 5V |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion