Learn More
Gilac™ Tote Caddy
Product Code. 17928481
Click to view available options
Color:
Orange
Yellow
Unit Size:
Each
Product Code. 17928481
Brand: Gilac™ G161428
Only null left
Add to basket
Add to basket
Tote Caddy is highly durable and suitable for universal use.
  • Highly durable.
  • Suitable for universal use.
  • Tote caddy can be used with 1 or 2 compartments.
Specifications
Color Yellow
Height (Metric) 205 mm
Length (Metric) 385 mm
Material HDPE
Type Utility Tray
Width (Metric) 255 mm
Includes 2 long compartments
Length (English) 15.2 in.
Height (English) 8.1 in.
Width (English) 10 in.
Autoclavable Not Autoclavable