Learn More
Ansell™ TouchNTuff™ 69-210 Series Natural Rubber Latex Gloves
Product Code. 15376957 Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Pack of 100
Product Code. 15376957
Brand: Ansell™ 69210070
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Powdered for easier donning and moisture absorption. Ansell TouchNTuff™ 69-210 Series Natural Rubber Latex Gloves offer superior tactile sensitivity for delicate tasks
  • Combined safety and comfort for delicate handling.
  • Ambidextrous, same glove fits both hands
  • Convenient and economical - stocks can be kept to a minimum
  • Flexible and comfortable to wear - cooler than vinyl, with superior finger flexibility and grip
  • Available in both powdered (69-210) and powder-free (69-318) versions
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Latex Free No
Material Rubber Latex
Powdered Powdered
Size Small
Thickness (Metric) 0.10 mm
Quantity 100/box
Series 69-210