Learn More
Ansell™ TouchNTuff™ 93-300 Series Green Nitrile Gloves
Complies with ASTM D 3577, Type II. Ansell™ TouchNTuff™ 93-300 Series Green Nitrile Gloves are ideal for use in electronics and semiconductor manufacturing, and laboratory testing.
Brand:  Ansell™ 93-300_6.5-7
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10441644
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • Delivers the highest protection for a wide range of chemicals than any other single use nitrile glove
  • Provides the wearer with a high level of comfort and increased protection in the forearm (longer cuff)
  • Manufactured from nitrile, offering three times the puncture resistance of standard latex gloves
  • Made without natural rubber latex proteins, the glove may be worn with confidence by latex sensitive (Type l) wearers
  • Suitable for ISO Class 5 or higher clean room environments
Specifications
Specifications
Nitrile
Nitrile
Yes
Gloves
93-300
Textured fingertips
240 Pairs
93-300
Standards CATEGORY III EN 374, EN 421, ASTM D 3577, Type II
Nitrile
Nitrile
6.5
0.12 mm
For blending of liquids, filtration processes, vaccines injectables operations etc.
Splash and high puncture resistance