Learn More
Motic™ Trace Comparison Microscopy Software
Use this software in conjunction with Motic™ Images Plus 2.0 to provide a comprehensive platform for evaluating compared images between microscopes. Motic™ Trace Comparison Microscopy Software allows you to perform a variety of comparative microscopy tasks.
Brand: Motic™ 1101001500722
Additional Details : Weight : 0.01000kg
Description
- Connect two or more Moticams or Motic™ Digital Microscopes to your computer and get immediate live comparison imaging.
- Cost effective.
- Overlay several live images, resize and independently rotate each image.
- Match two fingerprints with ease from live images or recall from a saved set.
- Check handwriting authenticity in a matter of seconds or determine whether two pieces of fabric belong to the same set.
Specifications
Trace Comparison Microscopy | |
Comparison microscopy, forensics |