Learn More
Alla France™ Traceable Mineral Oil Hydrometer
Product Code. 11762565
Click to view available options
Specific Gravity Range:
29 to 41° API
9 to 21° API
Unit Size:
Each
Product Code. 11762565
Brand: Alla France™ 3400FC009/60qp
Only null left
Specifications
Quantity 1Each
Type Traceable Mineral Oil Hydrometer
Accuracy ±0.1°API
Length (Metric) 335 mm
Baume Range 9 to 21°API
Specific Gravity Range 9 to 21° API