Learn More
Alla France™ Traceable Mineral Oil Hydrometer
Brand: Alla France™ 3400FC029/60-qp
Additional Details : Weight : 0.80000kg
Specifications
1Each | |
±0.1°API | |
29 to 41°API |
Traceable Mineral Oil Hydrometer | |
335 mm | |
29 to 41° API |