missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Stick Thermometer

Recall the highest or lowest temperatures achieved over any time period with the touch of a button. Fisherbrand™ Traceable™ Stick Thermometers simultaneously display probe and ambient temperatures.

Brand:  Fisherbrand™

Additional Details : Weight : 1.00000kg

Product Code. 12764088

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »


Description

Description

v

  • Minimum/maximum memory thermometer measures liquid, air/gas, or semisolids to a resolution of 0.1°
  • Range is –58.0 to 158.0°F or –50 to 70°C with an accuracy of ±1°C
  • Multi-point calibration on an individually-numbered Traceable Certificate which assures accuracy from our ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA.  
  • Indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement
  • Stick Thermometer’s submersible sensor dimensions are 0.2 inch diameter by 5/8 inch length and submersible cable length is 3 feet
  • One ounce weight makes it perfect for carrying in the pocket, or it may be placed anywhere using the supplied self-adhesive Velcro tabs
  • Furnished silver-oxide battery powers unit continuously for a year
  • Chemical-resistant and shockproof ABS plastic case is 4-/4 x 1 x 5/8 inches
Specifications

Specifications

Traceable™ Certificate is provided from our ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA
3 ft. Cable, Probe with Removable Suction Cup, Velcro™ Tabs, Battery
0.1°C
70°C
-50°C to +70°C
5 mm
15.87 mm
Stick Thermometer
-50°C
±1°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions