Learn More
Thermo Scientific™ TraceGOLD TG-200MS GC Columns
Product Code. 13478018 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
10 m
20 m
Unit Size:
Each
Product Code. 13478018
Brand: Thermo Scientific™ 260842750
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Exceptionally inert mid-polarity columns with selectivity and elution order optimized for difficult separations
  • Polar phase, trifluoropropylmethyl polysiloxane solid phase resolves compounds that phenyl and cyano phases cannot
  • Outstanding thermal stability and low bleed
  • Suitable for use with sensitive detectors including ECDs, NPDs and MS
  • Confirmation column in combination with another GC column
  • Equivalent to USP G6 phase
  • Similar to Rtx™-200MS, DB-200, DB-210
TRUSTED_SUSTAINABILITY
Specifications
Max. Temperature -20°C to +320/340°C
Length (English) Capillary 32.8 ft.
Type GC Capillary Column
Diameter (Metric) Inner 0.15 mm
For Use With (Application) Silica Analysis
Film Thickness 0.15 μm
Length (Metric) 10 m