missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Aminomethyl-1-cyclohexanol, 99+%

Product Code. p-57443 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
2.5 g
Packaging:
Glass bottle
Unit Size:
1g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10592641 1 g Glass bottle 1g
10359340 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10592641 Supplier Thermo Scientific Acros Supplier No. 265890010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 5691-09-8 | C7H15NO | 129.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5691-09-8
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.2
MDL Number MFCD06213163
InChI Key JGKFBZBVCAWDFD-NKWVEPMBSA-O
Synonym trans-2-aminomethyl-1-cyclohexanol, trans-2-aminomethyl-cyclohexanol, 1r,2s-2-aminomethyl cyclohexan-1-ol, jgkfbzbvcawdfd-nkwvepmbsa-n, 1r,2s-2-aminomethyl cyclohexanol, 1r,2s---trans-2-aminomethyl cyclohexanol, trans-2-aminomethyl cyclohexanol, 2-aminomethyl cyclohexanol, e, cyclohexanol, 2-aminomethyl-, trans, 1r-2alpha-aminomethylcyclohexan-1beta-ol
PubChem CID 2724658

Specifications

Color Colorless
Boiling Point 226°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula OHC6H10CH2NH2
Refractive Index 1.4905 to 1.4925
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform,ethanol,methanol
Formula Weight 129.2
Percent Purity 99+%
Physical Form Liquid After Melting
Chemical Name or Material trans-2-Aminomethyl-1-cyclohexanol, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • trans-2-Aminomethyl-1-cyclohexanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.