missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexenoic acid, 96%, Thermo Scientific Chemicals

Catalog Number p-7021293
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13419-69-7
C6H9O2
113.14
MFCD00002705
NIONDZDPPYHYKY-SNAWJCMRSA-M
trans-2-hexenoic acid, 2-hexenoic acid, e-hex-2-enoic acid, hex-2-enoic acid, trans-hex-2-enoic acid, e-2-hexenoic acid, hexenoic acid, 2e-hex-2-enoic acid, 2-hexenoic acid, 2e, 2-hexenoic acid, e
5282707
CHEBI:87721
(E)-hex-2-enoic acid
CCC\C=C\C([O-])=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13419-69-7
C6H9O2
113.14
MFCD00002705
NIONDZDPPYHYKY-SNAWJCMRSA-M
trans-2-hexenoic acid, 2-hexenoic acid, e-hex-2-enoic acid, hex-2-enoic acid, trans-hex-2-enoic acid, e-2-hexenoic acid, hexenoic acid, 2e-hex-2-enoic acid, 2-hexenoic acid, 2e, 2-hexenoic acid, e
5282707
CHEBI:87721
(E)-hex-2-enoic acid
CCC\C=C\C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 30°C to 35°C
Density 0.965
Boiling Point 217°C
Flash Point 125°C (257°F)
Refractive Index 1.4385
Quantity 10 g
UN Number UN3261
Beilstein 1720443
Formula Weight 114.14
Percent Purity 96%
Chemical Name or Material trans-2-Hexenoic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'