missing translation for 'onlineSavingsMsg'
Learn More

trans-2-[trans-2-(1,3,3-Trimethyl-2-indolinylidene)ethylidene]-1-indanone, 96%, Alfa Aesar™

Brand:  Alfa Aesar H31932.06

Additional Details : CAS : 53704-23-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15407945

  • 207.00€ / 5g


Estimated Shipment
27-04-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

53704-23-7
315.416
RMZNRQVMHQVFHH-UHFFFAOYSA-N
53426998
CC1(C2=CC=CC=C2N(C1=CC=C3CC4=CC=CC=C4C3=O)C)C
C22H21NO
MFCD06409584
2e-2-2-2e-1,3,3-trimethylindol-2-ylidene ethylidene-3h-inden-1-one, 2e-2-2e-2-1,3,3-trimethyl-1,3-dihydro-2h-indol-2-ylidene ethylidene indan-1-one, e-2-2-e-1,3,3-trimethylindolin-2-ylidene ethylidene-2,3-dihydro-1h-inden-1-one, trans-2-trans-2-1,3,3-trimethyl-2-indolinylidene ethylidene-1-indanone, e-2-e-2-1,3,3-trimethylindolin-2-ylidene ethylidene-2,3-dihydro-1h-inden-1-one, 2-2-1,3,3-trimethylindolin-2-ylidene ethylidene-2,3-dihydro-1h-inden-1-one
2-[2-(1,3,3-trimethylindol-2-ylidene)ethylidene]-3H-inden-1-one

Specifications

trans-2-[trans-2-(1,3,3-Trimethyl-2-indolinylidene)ethylidene]-1-indanone
5g
C22H21NO
2e-2-2-2e-1,3,3-trimethylindol-2-ylidene ethylidene-3h-inden-1-one, 2e-2-2e-2-1,3,3-trimethyl-1,3-dihydro-2h-indol-2-ylidene ethylidene indan-1-one, e-2-2-e-1,3,3-trimethylindolin-2-ylidene ethylidene-2,3-dihydro-1h-inden-1-one, trans-2-trans-2-1,3,3-trimethyl-2-indolinylidene ethylidene-1-indanone, e-2-e-2-1,3,3-trimethylindolin-2-ylidene ethylidene-2,3-dihydro-1h-inden-1-one, 2-2-1,3,3-trimethylindolin-2-ylidene ethylidene-2,3-dihydro-1h-inden-1-one
CC1(C2=CC=CC=C2N(C1=CC=C3CC4=CC=CC=C4C3=O)C)C
315.416
315.42
184°C to 187°C
53704-23-7
MFCD06409584
RMZNRQVMHQVFHH-UHFFFAOYSA-N
2-[2-(1,3,3-trimethylindol-2-ylidene)ethylidene]-3H-inden-1-one
53426998
96%
Videos
SDS Documents