Learn More
Alfa Aesar™ trans-4-(Boc-amino)cyclohexylamine, 97%
Brand: Alfa Aesar™ H62734.14
Additional Details : CAS : 177906-48-8 Weight : 0.02500kg
Quantity | 25g |
---|
Description
Applicationstrans-4-(Boc-amino)cyclohexylamine is used in the preparation of orally bioavailable and selective V1A receptor antagonist.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
177906-48-8 | |
214.309 | |
FEYLUKDSKVSMSZ-UHFFFAOYSA-N | |
2756050 | |
CC(C)(C)OC(=O)NC1CCC(CC1)N |
C11H22N2O2 | |
MFCD03001719 | |
trans-n-boc-1,4-cyclohexanediamine, n-boc-trans-1,4-cyclohexanediamine, 1-n-boc-cis-1,4-cyclohexyldiamine, tert-butyl 4-aminocyclohexyl carbamate, n-boc-1,4-cyclohexanediamine, tert-butyl n-4-aminocyclohexyl carbamate, tert-butyl cis-4-aminocyclohexyl carbamate, 1-boc-amino-1,4-cyclohexanediamine, tert-butyl 4-aminocyclohexylcarbamate, cis-n-boc-1,4-cyclohexanediamine | |
tert-butyl N-(4-aminocyclohexyl)carbamate |
Specifications
trans-4-(Boc-amino)cyclohexylamine | |
177906-48-8 | |
MFCD03001719 | |
Slightly soluble in water. | |
CC(C)(C)OC(=O)NC1CCC(CC1)N | |
214.309 | |
214.3 |
25g | |
C11H22N2O2 | |
trans-n-boc-1,4-cyclohexanediamine, n-boc-trans-1,4-cyclohexanediamine, 1-n-boc-cis-1,4-cyclohexyldiamine, tert-butyl 4-aminocyclohexyl carbamate, n-boc-1,4-cyclohexanediamine, tert-butyl n-4-aminocyclohexyl carbamate, tert-butyl cis-4-aminocyclohexyl carbamate, 1-boc-amino-1,4-cyclohexanediamine, tert-butyl 4-aminocyclohexylcarbamate, cis-n-boc-1,4-cyclohexanediamine | |
FEYLUKDSKVSMSZ-UHFFFAOYSA-N | |
tert-butyl N-(4-aminocyclohexyl)carbamate | |
2756050 | |
97% |