missing translation for 'onlineSavingsMsg'
Learn More

(+)-trans Taxifolin, TRC

Product Code. 30409379
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30409379

Brand: Toronto Research Chemicals TRCT010005100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (+)-Taxifolin
CAS 480-18-2
Molecular Formula C15 H12 O7
Quantity 100 mg
Synonym 4H-1-Benzopyran-4-one','2-(3,4-dihydroxyphenyl)-2,3-dihydro-3,5,7-trihydroxy-','(2R,3R)-, 4H-1-Benzopyran-4-one','2-(3,4-dihydroxyphenyl)-2,3-dihydro-3,5,7-trihydroxy-','(2R-trans)-, Flavanone','3,3',4',5,7-pentahydroxy- (8CI), (2R,3R)-2-(3,4-Dihydroxyphenyl)-2,3-dihydro-3,5,7-trihydroxy-4H-1-benzopyran-4-one, (+)-(2R,3R)-Dihydroquercetin, (+)-Dihydroquercetin, (+)-Taxifolin, (+)-trans-Taxifolin, (2R,3R)-(+)-Taxifolin, (2R,3R)-3,3',4',5,7-Pentahydroxyflavanone, (2R,3R)-Dihydroquercetin, 2,3-Dihydroquercetin, 3,5,7,3',4'-Pentahydroxyflavanone, Dihydroquercetin, Diquertin, Distylin, Flamena, Flamena D, Jikuberuchin, Lariksin, Lavitol, Taxifolin, Taxifoliol
InChI Formula InChI=1S/C15H12O7/c16-7-4-10(19)12-11(5-7)22-15(14(21)13(12)20)6-1-2-8(17)9(18)3-6/h1-5,14-19,21H/t14-,15+/m0/s1
SMILES O[C@@H]1[C@H](Oc2cc(O)cc(O)c2C1=O)c3ccc(O)c(O)c3
IUPAC Name (2R,3R)-2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-2,3-dihydrochromen-4-one
Molecular Weight (g/mol) 304.25
Formula Weight 304.0583
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.