missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-2,4-Hexadien-1-ol, 98%, stab. with 0.1% alpha-tocopherol, Thermo Scientific™

Product Code. 11480523
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11480523

Brand: Thermo Scientific Alfa Aesar A12660.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Trans,trans-2,4-Hexadien-1-ol is used in the preparation of trans-2,3-methano-4(Z)-hexenol by reacting with diiodomethane. It is used as a flavor and fragrance agent having pineapple odor.

Solubility
Slightly miscible with water.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17102-64-6
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00002925
InChI Key MEIRRNXMZYDVDW-MQQKCMAXSA-N
Synonym sorbic alcohol, 2,4-hexadien-1-ol, sorbyl alcohol, trans,trans-2,4-hexadien-1-ol, 2,4-hexadienol, hexacose, hexakose, sorbinic alcohol, 1-hydroxy-2,4-hexadiene, 2e,4e-hexa-2,4-dien-1-ol
PubChem CID 641256
IUPAC Name (2E,4E)-hexa-2,4-dien-1-ol
SMILES C\C=C\C=C\CO

Specifications

Melting Point 30°C to 31°C
Boiling Point 79°C to 80°C (12 mmHg)
Flash Point 72°C (161°F)
Quantity 5 g
Beilstein 1719718
Sensitivity Air and light sensitive
Solubility Information Slightly miscible with water.
Formula Weight 98.15
Percent Purity 98%
Chemical Name or Material trans,trans-2,4-Hexadien-1-ol, stabilized with 0.1% alpha-tocopherol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.