Learn More
Neogen Transfer Pipette TP100
Product Code. 17208550
Encompass
Click to view available options
:
Quantity: 100 Pk.
Product Code. 17208550
Brand: Neogen 7100177474
This item requires a price update prior to being ordered.
Request a Price for this item here.
Intended for use with the Neogen Allergen Protein Rapid Kits to transfer clean-in-place (CIP) final rinse water to the Neogen Extraction Buffer, or any extracted sample to the Neogen Allergen Protein Lateral Flow Device (LFD).
  • 100 μL transfer pipette
  • Pipette designed for use with Neogen Allergenic Protein Rapid Detection Test Kits
  • 100 per pack
Specifications
For Use With (Application) Neogen Allergenic Protein Rapid Detection Test Kits
Quantity 100 Pk.
Volume (Metric) 100 μL