Learn More
enQuireBio™ TranslationBlocker Human IGF1R siRNA
Blocks transcription and translation of IGF1R from Human.
Brand:  enQuireBio™ QX46-1nmol
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16029241
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Specifications
NP_000866
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
3480
1 nmol
Human
ME Human Melanoma Cells, MDA-MB-231 Human Breast Cancer Cells, and UC101 Ovarian Cancer Cells.
IGF1R
siRNA
JTK13, CD221, IGFIR, IGFR, Tyrosine-protein kinase receptor Insulin-like growth factor 1 receptor
Lyophilized