Learn More
BRAND™ Translucent Polypropylene T-shape Tubing Connector
Constructed of translucent polypropylene. Accommodates three tubes with T-style side arm.
Brand:  BRAND™ 152800
Additional Details : Poids : 0.06000kg
 View more versions of this product
Product Code. 15241509
  • 16.20€ / Pack of 20
Expédition estimée
01-10-2020
Add to basket
Description
Description
  • Translucent polypropylene construction
  • Accommodates 3 tubes of the same diameter
Specifications
Specifications
Translucent
20 mm
20 Pack
T-shape
Polypropylene
Tubing Connector
3 to 4 mm tubing I.D.