Learn More
Rogo Sampaic™ Transparent Glass Coverslips
Product Code. 14872231
Click to view available options
Dimensions (L x W):
20 x 20 mm
22 x 22 mm
Unit Size:
Pack of 100
Pack of 1000
Product Code. 14872231
Brand: Rogo Sampaic™ BPD003
Only null left
Add to basket
Add to basket
Glass coverslips are transparent and don't leave fingerprints.
Glass coverslips are transparent and don't leave fingerprints.
  • Manufactured in soda glass
  • Uniform thickness of 0.13 to 0.16 mm
Specifications
Shape Square
Material Soda Lime Glass
Thickness (Metric) #1 (0.13 to 0.16 mm)
Dimensions (L x W) 20 x 20 mm
Description Glass Coverslip
Quantity 100