Learn More
Carl Zeiss™ Transport Case for Stemi 305/508 (D)
Click to view available options
For Use With (Equipment):
Stemi 305/508, stand K, front optics and illuminators K
Unit Size:
Each
Description
- Trolley Case for transport and storage for Stemi 305/508, stand K, front optics and illuminators K
- Lateral handles, lockable and dustproof
- Suitable for shipment only with additional packaging
- Outer dimensions 615 x 540 x 325mm
- weight 8kg
Specifications
Specifications
| Description | Lateral handles, lockable and dustproof |
| Product Type | Transport Case for Stemi 305/508 (D) |
| For Use With (Equipment) | Stemi 305/508, stand K, front optics and illuminators K |
| Dimensions (L x W x H) | 615 x 540 x 325 mm |
| Weight (Metric) | 8 kg |