missing translation for 'onlineSavingsMsg'
Learn More

Tri-Sodium Orthophosphate Dodecahydrate, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7073554
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
10101-89-0
H24Na3O16P
380.119
149198
ASTWEMOBIXQPPV-UHFFFAOYSA-K
trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate
61473
trisodium;phosphate;dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]
This item is not returnable. View return policy
10101-89-0
H24Na3O16P
380.119
149198
ASTWEMOBIXQPPV-UHFFFAOYSA-K
trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate
61473
trisodium;phosphate;dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]
Melting Point 75°C
Color White
pH 11.8 to 12
Physical Form Solid
Quantity 250 g
Formula Weight 380.13g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • tri-Sodium orthophosphate dodecahydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Tribasic sodium phosphate dodecahydrate,Trisodium phosphate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'