Learn More
Triacetoxy(methyl)silane, tech. 90%, Thermo Scientific Chemicals
Catalog Number 11436856
Click to view available options
Quantity:
10 g
50 g
250 g
4253-34-3
C7H12O6Si
220.25
MFCD00008694
TVJPBVNWVPUZBM-UHFFFAOYSA-N
methyltriacetoxysilane, methylsilanetriyl triacetate, triacetoxymethylsilane, triacetoxy methyl silane, silanetriol, methyl-, triacetate, silane, methyltriacetoxy, methylsilanetriol triacetate, methyltrihydroxysilane triacetate, apk 1 silane derivative, triacetoxy-methylsilane
77929
CC(=O)O[Si](C)(OC(C)=O)OC(C)=O
4253-34-3
C7H12O6Si
220.25
MFCD00008694
TVJPBVNWVPUZBM-UHFFFAOYSA-N
methyltriacetoxysilane, methylsilanetriyl triacetate, triacetoxymethylsilane, triacetoxy methyl silane, silanetriol, methyl-, triacetate, silane, methyltriacetoxy, methylsilanetriol triacetate, methyltrihydroxysilane triacetate, apk 1 silane derivative, triacetoxy-methylsilane
77929
CC(=O)O[Si](C)(OC(C)=O)OC(C)=O
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Boiling Point 87°C to 88°C (3mmHg)
Melting Point 40°C to 45°C
Quantity 10 g
UN Number UN1759
Beilstein 1788668
Flash Point 85°C (185°F)
IUPAC Name [diacetyloxy(methyl)silyl] acetate
Formula Weight 220.25
Percent Purity 96%
Sensitivity Moisture sensitive
Density 1.175
Chemical Name or Material Triacetoxy(methyl)silane