Learn More
Kartell™ Triangular Stirring Bars
Cuneiform. Ideal for dissolving solids and mixing sediments thanks to its scraping action.
Brand:  Kartell™ 0078300
Product Code. 10356251
  • ONLINE EXCLUSIVE PRICE
    81.00€
    66.63€ / Pack of 10
    Save 14.37€
    17% Off
Only null left
Estimated Shipment: 17-02-2026
Description
  • Angled surfaces can provide lots of turbulence even at slow speeds
  • The shape means they can act as a scraper, so they’re good for helping with dissolution or preventing sedimentation
  • Suitable for foodstuff
  • Product Suggestions