Learn More
BRAND™ Triangular Wedge-Shape PTFE Magnetic Stirring Bars
Product Code. 12005292 Shop All Brand Products
Change view
Click to view available options
Length (Metric):
25 mm
35 mm
40 mm
50 mm
55 mm
80 mm
Diameter (Metric):
8 mm
10 mm
12 mm
14 mm
16 mm
Unit Size:
Pack of 10
Product Code. Length (Metric) Diameter (Metric) unitSize
12005292 35 mm 10 mm Pack of 10
12095282 25 mm 8 mm Pack of 10
10782521 40 mm 14 mm Pack of 10
11854100 50 mm 12 mm Pack of 10
10668691 55 mm 14 mm Pack of 10
10039820 80 mm 16 mm Pack of 10
10762351 25 mm 14 mm Pack of 10
7 options
Product Code. 12005292
Brand: BRAND™ 137815
Only null left
Add to basket
Add to basket
An excellent option for liquid agitation in the laboratory.
  • Ideal for dissolving solids and mixing of sediments due to scraping action
  • Angular surface creates high turbulence even at low speeds
  • Provided in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
Description Triangular wedge-shaped
Diameter (Metric) 10 mm
Length (Metric) 35 mm
Material PTFE
Quantity 10/Pk.
Shape Triangle