missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Acid, SPEX CertiPrep™

Product Code. 15256256 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15256256 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15256256 Supplier SPEX Certiprep Supplier No. S3575

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Trichloroacetic acid76-03-91000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging 2mL Amber Prescored Glass Ampule
CAS 76-03-9, 1634-04-4
CAS Min % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molecular Formula C2HCl3O2
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
SMILES C(=O)(C(Cl)(Cl)Cl)O
IUPAC Name 2,2,2-trichloroacetic acid
Solvent or Matrix MTBE
Molecular Weight (g/mol) 163.378
PubChem CID 6421
Quantity 1 mL
ChEBI CHEBI:30956
Chemical Name or Material Trichloroacetic Acid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trichloroacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Methyl-tert-butyl ether, trichloroaceticacid

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.