Learn More
Honeywell™ TRICOTON Ultra Light Gloves
Honeywell™ TRICOTON Ultra Light Gloves are a 100% cotton lightweight knitted glove constructed of ultralight yarn.
Brand:  Honeywell™ RGT350T7
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 12693333
Quantity Price
1 1.16€ / Each
10 1.04€ / Each
In stock, ready to ship: 190
Add to basket
Description
Description
  • 100% cotton lightweight knitted glove
  • The knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
  • The glove offers maximum comfort and added sturdiness
  • Of a neutral pH level and without any treatment, this cotton is tolerated perfectly well by the skin and limits the absorption of perspiration
  • The tight knit gives the glove perfect support, limits the penetration of dirt, offers a good touch while offering protection against small cuts
  • Light color enhances the glove's hygiene and cleanliness properties
Specifications
Specifications
13 gauge
Cotton
230 mm
6 cm elastic wrist
7
EN 420-03 EEC Basic standards