missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, 97%, Thermo Scientific Chemicals

Product Code. 10145303 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
1 L
2.5 L
25 L
Unit Size:
1 L
100mL
2.5 L
25 L
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10145303

Brand: Thermo Scientific Acros 139561000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-71-6
Molecular Weight (g/mol) 149.19
MDL Number MFCD00002855
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
Synonym daltogen, triethanolamine, 2,2',2-nitrilotriethanol, thiofaco t-35, sterolamide, trihydroxytriethylamine, nitrilotriethanol, trolamine, tris 2-hydroxyethyl amine, triethylolamine
PubChem CID 7618
ChEBI CHEBI:28621

Specifications

Decomposition Information >325°C
Melting Point 21.0°C
Density 1.1245 g/mL
Boiling Point 360.0°C
Flash Point 190°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C6H15NO3
Linear Formula (HOCH2CH2)3N
Refractive Index 1.4840 to 1.4860
Quantity 100 mL
Fieser 01,1196
Merck Index 15, 9830
Specific Gravity 1.1245
Solubility Information Solubility in water: freely soluble. Other solubilities: miscible with alcohol,methanol and acetone,soluble in chloroform,slichtly soluble in benzene and ether,soluble 4.2% in benzene; 1.6% in ether; 0.4% in,carbon tetrachloride and < 0.1% in n-he
SMILES OCCN(CCO)CCO
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethan-1-ol
Viscosity 600 mPa.s (25°C)
Formula Weight 149.19
Vapor Pressure <0.01 mmHg (at 20°C)
Percent Purity 97%
Physical Form Oily Liquid
Chemical Name or Material Triethanolamine, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.