missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, Extra Pure, SLR, Fisher Chemical™

Product Code. 10458420 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
25 L
Packaging:
Amber glass bottle
Metal can
Unit Size:
2.5 L
25 L
500mL
Product Code. Quantity Packaging unitSize
10458420 500 mL Amber glass bottle 500mL
10152640 2.5 L Amber glass bottle 2.5 L
10224690 25 L Metal can 25 L
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10458420

Brand: Fisher Chemical T/3150/08

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Triethanolamine, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-71-6
Molecular Formula C6H15NO3
Molecular Weight (g/mol) 149.19
MDL Number 2855
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
Synonym triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
PubChem CID 7618
ChEBI CHEBI:28621
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethanol
SMILES C(CO)N(CCO)CCO

Specifications

Melting Point 21°C
Color Yellow
pH 10.5
Boiling Point 360°C
Packaging Amber glass bottle
Quantity 500 mL
Vapor Density 5.14
Formula Weight 149.19g/mol
Vapor Pressure <0.01mmHg at 20°C
Physical Form Viscous Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.