missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, TRC

Product Code. 30451923
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30451923 10 g 10g
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30451923

Brand: Toronto Research Chemicals TRCT77558010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trolamine
CAS 102-71-6
Molecular Formula C6 H15 N O3
Quantity 10 g
Synonym Trolamine, 2,2',2''-Nitrilotriethanol, 2,2',2''-Nitrilotrisethanol, Ethanol','2,2',2''-nitrilotri- (8CI), 2,2',2''-Nitrilotriethanol, 2,2',2''-Nitrilotris[ethanol], 2-[Bis(2-hydroxyethyl)amino]ethanol, Alkanolamine 244, Biafine, Daltogen, N,N,N-Tri(2-hydroxyethyl)amine, NSC 36718, Nitrilotriethanol, Rhenofit 3555, S 80, S 80 (amine), Sterolamide, Sting-Kill, TEA, TEA (amino alcohol), TEA 99LFG, TEOA, Tri(2-hydroxyethyl)amine, Triethanolamin, Triethanolamine, Tris(2-hydroxyethyl)amine, Tris(β-hydroxyethyl)amine, Tris-amino Ultra PC, Trolamine, tris-(2-Hydroxyethyl)amine
InChI Formula InChI=1S/C6H15NO3/c8-4-1-7(2-5-9)3-6-10/h8-10H,1-6H2
SMILES OCCN(CCO)CCO
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethanol
Molecular Weight (g/mol) 149.19
Formula Weight 149.1052
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.