missing translation for 'onlineSavingsMsg'
Learn More

Triethyl phosphite, 98%, Thermo Scientific Chemicals

Catalog Number 11380004
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
122-52-1
C6H15O3P
166.157
MFCD00009084
BDZBKCUKTQZUTL-UHFFFAOYSA-N
phosphorous acid, triethyl ester, triethoxyphosphine, ethyl phosphite, eto 3p, triethylphosphite, phosphorous acid triethyl ester, fosforyn trojetylowy czech, unii-6b2r04s55g, hsdb 895, p oet 3, c2h5o 3p
31215
triethyl phosphite
CCOP(OCC)OCC
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
122-52-1
C6H15O3P
166.157
MFCD00009084
BDZBKCUKTQZUTL-UHFFFAOYSA-N
phosphorous acid, triethyl ester, triethoxyphosphine, ethyl phosphite, eto 3p, triethylphosphite, phosphorous acid triethyl ester, fosforyn trojetylowy czech, unii-6b2r04s55g, hsdb 895, p oet 3, c2h5o 3p
31215
triethyl phosphite
CCOP(OCC)OCC

Triethyl phosphite is used as a ligand in organometallic chemistry and as a reagent in organic synthesis. It is used as reference to phosphoric acid standard in 31P NMR spectroscopy. It acts as a reducing agent to prepare 2-phenylindazole from o-nitrobenzylidineaniline as well as reacts with electrophiles to get phosphonates. It forms a stable complex with copper(I) iodide. It finds application in a modified Staudinger reaction for the alkyl bromide to an amine through the azide. Further, it is also used in the preparation of 3-phenyl-2-substituted indoles by reacting with beta-nitro styrenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl phosphite is used as a ligand in organometallic chemistry and as a reagent in organic synthesis. It is used as reference to phosphoric acid standard in 31P NMR spectroscopy. It acts as a reducing agent to prepare 2-phenylindazole from o-nitrobenzylidineaniline as well as reacts with electrophiles to get phosphonates. It forms a stable complex with copper(I) iodide. It finds application in a modified Staudinger reaction for the alkyl bromide to an amine through the azide. Further, it is also used in the preparation of 3-phenyl-2-substituted indoles by reacting with beta-nitro styrenes.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Melting Point -112°C
Density 0.958
Boiling Point 155°C to 157°C
Flash Point 43°C (109°F)
Odor Unpleasant
Refractive Index 1.413
Quantity 100 mL
UN Number UN2323
Beilstein 956578
Sensitivity Air and moisture sensitive
Solubility Information Miscible with alcohol and ether. Immiscible with water.
Formula Weight 166.16
Percent Purity 98%
Chemical Name or Material Triethyl phosphite
Show More Show Less
compliance-icons
Product Identifier
  • Triethyl phosphite
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : triethyl phosphite

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'