missing translation for 'onlineSavingsMsg'
Learn More

(Trifluoromethyl)trimethylsilane, 99%, Thermo Scientific Chemicals

Catalog Number p-204962
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
81290-20-2
C5H11F3Si
156.22
MFCD00145454
MTYSDPUZDZURPP-UHFFFAOYSA-N
trifluoromethyl trimethylsilane, trimethyl trifluoromethyl silane, trifluoromethyltrimethylsilane, ruppert's reagent, tms-cf3, tfmtms, silane, trimethyl trifluoromethyl, unii-a009786qpj, trimethylsilyl trifluoromethane, ruppert-prakash reagent
552549
trimethyl(trifluoromethyl)silane
CC(C)(C)[SiH2]C(F)(F)F
This item is not returnable. View return policy
81290-20-2
C5H11F3Si
156.22
MFCD00145454
MTYSDPUZDZURPP-UHFFFAOYSA-N
trifluoromethyl trimethylsilane, trimethyl trifluoromethyl silane, trifluoromethyltrimethylsilane, ruppert's reagent, tms-cf3, tfmtms, silane, trimethyl trifluoromethyl, unii-a009786qpj, trimethylsilyl trifluoromethane, ruppert-prakash reagent
552549
trimethyl(trifluoromethyl)silane
CC(C)(C)[SiH2]C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.9500g/mL
Boiling Point 54.0°C to 55.0°C
Flash Point -17°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3SiCF3
Refractive Index 1.3300 to 1.3320
Quantity 1 mL
Merck Index 15, 9851
Specific Gravity 0.95
Formula Weight 142.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (Trifluoromethyl)trimethylsilane, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (Trifluoromethyl)trimethylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'