missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluralin-(dipropyl-d14), TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Trifluralin D14 (di-n-propyl D14) |
| CAS | 347841-79-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C13 2H14 H2 F3 N3 O4 |
| Quantity | 5 mg |
| Synonym | Benzenamine','2,6-dinitro-N,N-di(propyl-d7)-4-(trifluoromethyl)- (9CI), 2,6-Dinitro-N,N-di(propyl-d7)-4-(trifluoromethyl)benzenamine, 2,6-Dinitro-N,N-di(propyl-d7)-4-(trifluoromethyl)aniline, Trifluralin-d14, Trifluralin D14 (di-n-propyl D14), Trifluralin-D14 (N,N-dipropyl-D14) |
| InChI Formula | InChI=1S/C13H16F3N3O4/c1-3-5-17(6-4-2)12-10(18(20)21)7-9(13(14,15)16)8-11(12)19(22)23/h7-8H,3-6H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])N(c1c(cc(cc1[N+](=O)[O-])C(F)(F)F)[N+](=O)[O-])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H] |
| IUPAC Name | N,N-bis(1,1,2,2,3,3,3-heptadeuteriopropyl)-2,6-dinitro-4-(trifluoromethyl)aniline |
| Molecular Weight (g/mol) | 349.37 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction