missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl borate, 99%, Thermo Scientific Chemicals

Catalog Number p-206851
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
121-43-7
MFCD00008346
WRECIMRULFAWHA-UHFFFAOYSA-N
trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
8470
CHEBI:38913
trimethyl borate
B(OC)(OC)OC
121-43-7
MFCD00008346
WRECIMRULFAWHA-UHFFFAOYSA-N
trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
8470
CHEBI:38913
trimethyl borate
B(OC)(OC)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -34.0°C
Color Colorless
Density 0.9150g/mL
Boiling Point 68.0°C to 69.0°C
Flash Point −8°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C3H9O3B
Linear Formula (CH3O)3B
Refractive Index 1.3562 to 1.3582
Quantity 100 g
Beilstein 01, 287
Fieser 01,673; 13,218
Merck Index 15, 9882
Specific Gravity 0.915
Solubility Information Solubility in water: reacts. Other solubilities: miscible with thf,diethyl ether,hexane,methanol,,isopropylamine,and other organic liquids
Molecular Weight (g/mol) 103.91
Viscosity 0.36 mPa.s (25°C)
Formula Weight 103.91
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Trimethyl borate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trimethyl borate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'