missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine (~30 wt. % Solution in Ethanol), TRC

Product Code. 30456439
Click to view available options
Quantity:
10 mL
25 mL
50 mL
Unit Size:
10mL
25mL
50mL
This item is not returnable. View return policy

Product Code. 30456439

Brand: Toronto Research Chemicals TRCT79167550ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethylamine
CAS 75-50-3
Molecular Formula C3 H9 N
Quantity 50 mL
Synonym Trimethylamine (8CI), N,N-Dimethylmethanamine, N,N,N-Trimethylamine, N-Trimethylamine, Methanamine, N,N-dimethyl-
InChI Formula InChI=1S/C3H9N/c1-4(2)3/h1-3H3
SMILES CN(C)C
IUPAC Name N,N-dimethylmethanamine
Molecular Weight (g/mol) 59.11
Formula Weight 59.0735
Concentration or Composition (by Analyte or Components) 30 %
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.