Learn More
Triphenylvinylsilane, 98%
Description
Triphenylvinylsilane can be used to produce triphenyl-[2-(tetrahydro-furan-2-yl)-ethyl]-silane at temperature of 190°C. This reaction will need reagent di(t-butyl)peroxide with reaction time of 2 hours. Also employed as an organic light emitting diode.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Triphenylvinylsilane can be used to produce triphenyl-[2-(tetrahydro-furan-2-yl)-ethyl]-silane at temperature of 190°C. This reaction will need reagent di(t-butyl)peroxide with reaction time of 2 hours. Also employed as an organic light emitting diode.
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture and heat. Store under inert gas.
Specifications
Specifications
| Melting Point | 68°C to 71°C |
| Quantity | 25 g |
| Linear Formula | (C6H5)3SiCH=CH2 |
| Beilstein | 913423 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 286.45 |
| Percent Purity | 98% |
| Sensitivity | Moisture sensitive |
| Chemical Name or Material | Triphenylvinylsilane |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.