Learn More
Ohaus™ Tripod Stand Base
OHAUS™ Tripod Stand Base provides secure support when working with a tripod.
Brand: Ohaus™ 30392363
Additional Details : Weight : 2.66400kg
Description
- OHAUS offers the largest selection of lab-frame kits, lab-lifts, rods and support stands in the industry to hold glassware safely
- Standard lab-frames or customisable set-ups with options to fit any space or application
- Select from 13 pre-configured lab-frame kits (connectors and feet included) to rods which can be purchased individually and combined with OHAUS accessories to build a customized frame
Specifications
Tripod Stand | |
Cast Iron | |
Clamp, Support, Tripod Stand, CLR-TBASEC | |
375 mm |