missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(4-bromophenyl)amine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Tris(4-bromophenyl)amine was used in the synthesis of porous luminescent covalent--organic polymers (COPs)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tris(4-bromophenyl)amine was used in the synthesis of porous luminescent covalent--organic polymers (COPs)
Solubility
Soluble in toluene.
Notes
Incompatible with strong oxidizing agents. Store under room temperature.
Tris(4-bromophenyl)amine was used in the synthesis of porous luminescent covalent--organic polymers (COPs)
Solubility
Soluble in toluene.
Notes
Incompatible with strong oxidizing agents. Store under room temperature.
Specifications
Melting Point | 141°C to 143°C |
Quantity | 10 g |
Solubility Information | Soluble in toluene. |
Formula Weight | 482.01 |
Percent Purity | 98% |
Chemical Name or Material | Tris(4-bromophenyl)amine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'