missing translation for 'onlineSavingsMsg'
Learn More

Merck Tris Buffer

Product Code. 10740113
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 10740113

Brand: Merck LSKCTB500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Requires

Requires Montage Plasmid 96 Miniprep Kit, Sold Separately

The Montage Plasmid MiniprepHTS 96 kit is a fast, easy to use kit for high purity plasmid or bacterial artificial chromosome (BAC) minipreps. Requires Montage Plasmid 96 Miniprep Kit, sold separately.

  • Fast, easy to use
  • For high purity plasmid or BAC minipreps
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris Buffer
Color Clear
Description Tris Buffer for Plasmid Miniprep and BAC Prep
Physical Form Solution
Quantity 500 mL
Solubility Information 20°C (Water)
Certifications/Compliance ISO 9001
Percent Purity MQ100, for non-regulated applications with no change notification requirements.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.