missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris Molecular Biology MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
Specifications
Specifications
| Quantity | 100 g |
| Formula Weight | 121.14 |
| Grade | Molecular Biology |
| Chemical Name or Material | Tris |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction