missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIS, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
250 g
5 kg
Unit Size:
250g
5kg
Description
- Tris has been useful as buffers in a wide variety of biological systems.
- Uses include pH control in vitro and in vivo for body fluids and in buffering systems for electrophoresis applications.
- Tris has been used as a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines (with aldehydes).
- Tris does not precipitate calcium salts and is of value in maintaining solubility of manganese salts.
- It can be used for the direct standardization of a strong acid solution; the equivalence point can be determined either potentiometrically or by use of a suitable indicator such as 3-(4-Dimethylamino-1-naphthylazo)-4-methoxybenzenesulfonic acid.
- It is an auxiliary material in pharmaceutical science
Specifications
Specifications
| CAS | 77-86-1 |
| Melting Point | 167°C to 172°C |
| Boiling Point | 219°C to 220°C (at 10 mmHg) (Literature) |
| Color | White |
| pH | 10 to 11.5 (5% aq. soln.) |
| Physical Form | Crystalline Powder |
| Quantity | 250 g |
| Purity Grade Notes | ≥99.95% |
| Loss on Drying | ≤1% |
| Molecular Formula | C4H11NO3 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction