missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(trimethylsiloxy)silane, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Tris(trimethylsiloxy)silane |
| CAS | 1873-89-8 |
| Molecular Formula | C9H28O3Si4 |
| Quantity | 1 g |
| Synonym | 1,1,1,5,5,5-Hexamethyl-3-[(trimethylsilyl)oxy]trisiloxane, S 1003TS |
| InChI Formula | InChI=1S/C9H28O3Si4/c1-14(2,3)10-13(11-15(4,5)6)12-16(7,8)9/h13H,1-9H3 |
| SMILES | C[Si](C)(O[SiH](O[Si](C)(C)C)O[Si](C)(C)C)C |
| IUPAC Name | bis(trimethylsilyloxy)silyloxy-trimethylsilane |
| Molecular Weight (g/mol) | 296.66 |
| Formula Weight | 296.1116 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction