Learn More
Smeg™ Trolley System For Washing Pipettes
Used for washing volumetric pipettes up to 55cm in length. Smeg Instruments Trolley System for washing Pipettes has 20 positions for washing pipettes and/or flasks of 45 to 49cm in height.
Brand:  Smeg™ LPV40
Additional Details : Weight : 0.80000kg
Product Code. 12693860
  • 1820.00€ / Each
Estimated Shipment: 06-06-2024
to see stock.
Add to basket
Description
Description
  • Made of stainless steel trolley
  • 20 positions for pipettes of 55cm in height
  • 20 positions for pipettes and/or flasks of 45 to 49cm in height
Specifications
Specifications
Trolley System for Washing Pipettes