Learn More
Kimberly-Clark™ KLEENGUARD™ A50 Breathable Splash and Particle Protection Trousers
Specially designed for comfort, safety and strength. Kimberly-Clark™ KLEENGUARD™ A50 Breathable Splash and Particle Protection Trousers is designed for comfort, safety and general protection against larger particles, radioactive dust and chemical splashes.
Brand:  Kimberly-Clark™ 99500
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12671446
  • 114.00€ / Pack of 15
Estimated Shipment
02-03-2021
Add to basket
Description
Description
  • Approved as Complex Design (Category 3) equipment - offering protection to the levels specified for Type 5 and 6 by CEN
  • Single use and breathable protective trousers designed for maximum comfort and safety, providing anti-static and general protection against water-based or chemical splashes, liquids, and dust particles
  • Durable SMS fabric with additional treatment for increased protection
  • Suitable for use as protection against solid particles and particulate radioactive contamination - EN1073-2
  • Silicone-free, ultra-low lint and treated to be antistatic - to EN 1149-5
  • Ideal for protecting against powders, larger fibres and protection against a wide range of chemicals - reducing contamination in critical areas
  • Suitable for use  in pharmaceutical industries, manufacturing, utilities, electronics, agriculture and paint spraying areas
Specifications
Specifications
White
While working in chemical and pharmaceutical industries.
Double-layer fabric
Medium
Larger particles, radioactive dust and chemical splashes
Trousers