Learn More
Heathrow Scientific™ True North™ Mini-Cooler
Product Code. p-7193579 Shop All Thermo Scientific Incubateurs à CO2 Products
Click to view available options
Array:
3 x 4
4 x 8
8 x 12
Color:
Blue
Green
Description:
Clear Lid
Gel Filled Lid
Withstand Temperature (Metric):
-20°C
0°C
Unit Size:
Each
Product Code. 16390676
Brand: Heathrow Scientific™ HS120070
Only null left
Add to basket
Add to basket
The True North™ Mini Cooler protects temperature-sensitive samples at the bench or in the freezer. The Mini Cooler is ideal for keeping PCR™ reagents, enzymes, and other biochemical reagents cool while working at the bench.
  • Polycarbonate construction
  • Cooler has rubber non-slip feet
  • Gridded and keyed one-position lid for easy sample identification
  • Available with lid filled with insulating gel
Specifications
Color Green
Height (Metric) 10 cm
Height (English) 3.9 in.
Length (Metric) 14.5 cm
Length (English) 5.7 in.
Width (Metric) 10.5 cm
Width (English) 4.1 in.
Description Clear Lid
For Use With (Application) Tubes
Holds Tubes
Quantity 1 Ea.
Array 3 x 4
Material Polycarbonate
Type Cooler
Capacity (Metric) 0.5 to 2 mL
Withstand Temperature (Metric) 0°C