Learn More
Thermo Scientific™ Tryptone Soya Broth (USP) (Tube)
Description
Grow a wide variety of microorganisms with Thermo Scientific™ Oxoid™ Tryptone Soya Broth ready-prepared medium. The general purpose medium, containing two peptones, is suitable for the cultivation of both aerobes and anaerobes. It is specified in International Pharmacopoeia1,2,3,4and food testing methods for the growth of a wide variety of organisms.
Remove the need for autoclaving and dispensing with convenient, pre-filled and ready prepared tubes of Tryptone Soya Broth.
- Highly nutritious medium to support wide variety of microorganisms.
- Conforms to formulations detailed in International Pharmacopoeia1,2,3,4.
- Convenient: the pre-filled tubes are ready for immediate use.
- Simple: easy-to-use with minimum training.
- Long shelf life: up to 12 months
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Color | Maize yellow, transparent |
| Content And Storage | 2 to 25°C |
| Description | Tryptone Soya Broth (Tube) |
| Format | Poured tube |
| Manufacturing Origin | Germany |
| Product Type | Microbiology Media |
| Packaging Type | 50 tubes in a box |
| Quantity | 50/Pk. |