missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tryptose Phosphate Broth (Dehydrated)

Product Code. 11983762
Click to view available options
Quantity:
500 g
5 kg
Unit Size:
500g
5kg
This item is not returnable. View return policy

Product Code. 11983762

Brand: Thermo Scientific™ CM0283T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cultivate fastidious bacteria and supplement cell culture media with this medium.

Cultivate fastidious bacteria with Thermo Scientific™ Oxoid™ Tryptose Phosphate Broth (Dehydrated). Tryptose Phosphate Broth is recommended for the cultivation of streptococci, pneumococci, meningococci and other fastidious organisms. Tryptose Phosphate Broth with added agar and sodium azide is recommended for the isolation of pathogenic streptococci from cheese and other dairy products1,2.

Use Tryptone Phosphate Broth for cultivation of fastidious bacteria.

Not all products are available for sale in all territories. Please inquire

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: FDA.
Form Powder
Product Type Dehydrated Culture Media
Quantity 5 kg

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.