Learn More
BOLA Tube End Fittings
Product Code. p-200110022
Click to view available options
Color:
Black
Blue
Green
Orange
Red
Violet
White
Yellow
Diameter (Metric) Outer:
1.6 mm
1.61 mm
3.2 mm
Unit Size:
Pack of 10
Product Code. 10593822
Brand: BOLA F70206
Only null left
Add to basket
Add to basket
Tube End Fittings with Washers
  • Tube end fittings.
  • With male thread UNF ¼ in. 28 G washers made of PA are included in delivery.
  • Packing unit: 10 pieces, differing ordering quantities are rounded up to factor 10.
Specifications
Color White
Diameter (Metric) Outer 1.61 mm
Diameter (Metric) Inner 0.8 mm